missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)azobenzene, TRC

Product Code. 30377787
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377787 500 mg 500mg
30383111 5 g -
30380883 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377787 Supplier Toronto Research Chemicals Supplier No. TRCD495010500MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2481-94-9
Molecular Formula C16 H19 N3
Molecular Weight (g/mol) 253.34
Synonym Aniline, N,N-diethyl-p-(phenylazo)- (6CI,7CI), Benzenamine, N,N-diethyl-4-(phenylazo)- (9CI), C.I. Solvent Yellow 56 (8CI), N,N-Diethyl-4-(2-phenyldiazenyl)benzenamine, 4-(Diethylamino)azobenzene, 4-(N,N-Diethylamino)azobenzene, 4-(Phenylazo)-N,N-diethylaniline, Aizen SOT Yellow 1, Akrodye Oil Yellow 6, C.I. 11021, Ceres Yellow GGN, Diethyl yellow, Fast Oil Yellow 64403, Fat Yellow GGN, N,N-Diethyl-4-(phenylazo)aniline, N,N-Diethyl-4-aminoazobenzene, NSC 102374, Oil Yellow 2635, Oil Yellow DE, Oil Yellow DEA, Oil Yellow ENC, Oil Yellow GA, Oil Yellow GE, Oil Yellow NB, Oil Yellow SS Special, Oilsol Yellow DEA, Orient Oil Yellow 56, Orient Oil Yellow GGS, SOT Yellow 1, Sico Fat Yellow P, Solvent Yellow 56, Sudan Yellow 150, Sudan Yellow GGN, Waxoline Yellow ED, p-(Diethylamino)azobenzene
IUPAC Name N,N-diethyl-4-[(E)-phenyldiazenyl]aniline
SMILES CCN(CC)c1ccc(cc1)N=Nc2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H19N3/c1-3-19(4-2)16-12-10-15(11-13-16)18-17-14-8-6-5-7-9-14/h5-13H,3-4H2,1-2H3/b18-17+
Formula Weight 253.1579
Chemical Name or Material 4-(Diethylamino)azobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.