missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)salicylaldehyde, TRC

Product Code. 30386516
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386516 Supplier Toronto Research Chemicals Supplier No. TRCD4431855G
 Restricted Item
Estimated Shipment: 10-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material 4-(Diethylamino)salicylaldehyde
CAS 17754-90-4
Molecular Formula C11H15NO2
Quantity 5 g
Synonym 4-(Diethylamino)-salicylaldehyde, 4-(Diethylamino)-2-hydroxybenzaldehyde, 4-(N,N-Diethylamino)-2-hydroxybenzaldehyde, 4-(N,N-Diethylamino)salicylic Aldehyde, p-(Diethylamino)salicylaldehyde
InChI Formula InChI=1S/C11H15NO2/c1-3-12(4-2)10-6-5-9(8-13)11(14)7-10/h5-8,14H,3-4H2,1-2H3
SMILES CCN(CC)c1ccc(C=O)c(O)c1
Molecular Weight (g/mol) 193.24
Formula Weight 193.1103
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.