missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one, 97%, Thermo Scientific™

Product Code. 10571312
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571312 10 g Amber Glass Bottle 10g
10293222 1 g Amber Glass Bottle 1g
10452454 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571312 Supplier Thermo Scientific Maybridge Supplier No. RF03845EA
 Restricted Item
Estimated Shipment: 11-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67751-23-9
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.212
InChI Key DFZIBCAWOSFLFR-AATRIKPKSA-N
Synonym 4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, 1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, 1e-4,4-dimethoxy-3-oxobut-1-en-1-yl dimethylamine, 3e-4-dimethylamino-1,1-dimethoxybut-3-en-2-one, e-1,1-dimethoxy-4-dimethylaminobut-3-en-2-one, hcvl@nbfryyuffjjafp, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2, 4-dimethylamino-1,1-dimethoxy-3-butene-2-one, e-4-dimethylamino-1,1-dimethoxy-but-3-en-2-one
PubChem CID 5709580
IUPAC Name (E)-4-(dimethylamino)-1,1-dimethoxybut-3-en-2-one
SMILES CN(C)C=CC(=O)C(OC)OC

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)2NCH=CHC(O)CH(OCH3)2
Quantity 10 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.