missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylamino Antipyrine, TRC

Product Code. 30134824
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134824 1 g 1g
30137312 500 mg 500mg
30004232 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134824 Supplier Toronto Research Chemicals Supplier No. TRCD4608901G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-15-1
Molecular Formula C13 H17 N3 O
Molecular Weight (g/mol) 231.29
Synonym 3-Pyrazolin-5-one, 4-(dimethylamino)-2,3-dimethyl-1-phenyl- (7CI), Antipyrine, 4-(dimethylamino)- (8CI), 4-(Dimethylamino)-1,2-dihydro-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one, (Dimethylamino)analgesine, (Dimethylamino)antipyrine, (Dimethylamino)phenazone, 1,5-Dimethyl-4-dimethylamino-2-phenyl-3-pyrazolone, 1-Phenyl-2,3-dimethyl-4-dimethylaminopyrazol-5-one, 1-Phenyl-2,3-dimethyl-4-dimethylaminopyrazolone-5, 2,3-Dimethyl-4-dimethylamino-1-phenyl-3-pyrazolin-5-one, 2,3-Dimethyl-4-dimethylamino-1-phenyl-5-pyrazolone, 3-Keto-1,5-dimethyl-4-dimethylamino-2-phenyl-2,3-dihydropyrazole, 4-(Dimethylamino)antipyrine, 4-Dimethylamino-1-phenyl-2,3-dimethylpyrazolone, 4-Dimethylamino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, 4-Dimethylamino-2,3-dimethyl-1-phenyl-5-pyrazolone, 4-Dimethylaminophenazone, 4-N,N-Dimethylaminoantipyrine, Amidazophen, Amidazophene, Amidofebrin, Amidofen, Amidophen, Amidophenazone, Amidopyrazoline, Amidopyrin, Amidopyrine, Aminophenazon, Aminophenazone, Aminopyrin, Aminopyrine, Anafebrina, Brufaneuxol, Dereuma, Dimapyrin, Dimethylaminoazophene, Dimethylaminophenyldimethylpyrazone, Dipirin, Dipyrin, Dipyrine, Febrinina, Febron, Itamidone, Mamallet A, NSC 4993, Netsusarin, Novamidon, Piramidon, Piramidone, Piridol, Piromidina, Polinalin, Pyradone, Pyramidon, Pyramidone
IUPAC Name 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN(C)C1=C(C)N(C)N(C1=O)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C13H17N3O/c1-10-12(14(2)3)13(17)16(15(10)4)11-8-6-5-7-9-11/h5-9H,1-4H3
Formula Weight 231.1372
Chemical Name or Material Aminophenazone (4-Dimethylamino-1,5-dimethyl-2-phenyl-1,2-dihydro-3H-pyrazol-3-one)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.