missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzeneboronic acid, 97%, Thermo Scientific™

Product Code. 15492048
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15492048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15492048 Supplier Thermo Scientific Alfa Aesar Supplier No. H55723.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-(Dimethylamino)phenylboronic acid is a useful reagent for Suzuki-Miyaura cross-coupling reactions, Rhodium (Rh)-catalyzed asymmetric addition reactions, Palladium (Pd)-catalyzed C-OH bond activation. This reagent is used in Preparation of Push-pull arylvinyldiazine chromophores, Helical ortho-phenylene oligomers with terminal push-pull substitution, Alginate-supported cation-Pd nanoparticle gels as catalysts for the Suzuki-Miyaura cross-coupling reaction, Organic dyes for dye-sensitized solar cells.

Solubility
Sparingly soluble in water. (∼2.5%)

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28611-39-4
Molecular Formula C8H12BNO2
Molecular Weight (g/mol) 165.00
MDL Number MFCD01074642
InChI Key RIIPFHVHLXPMHQ-UHFFFAOYSA-N
Synonym 4-dimethylamino phenylboronic acid, 4-dimethylamino phenyl boronic acid, 4-n,n-dimethylamino phenylboronic acid, 4-dimethylamino benzeneboronic acid, 4-dimethylaminophenylboronic acid, 4-dimethylaminophenyl boronic acid, 4-dimethylamino phenyl boranediol, 4-n,n-dimethylphenylboronic acid
PubChem CID 2734344
IUPAC Name [4-(dimethylamino)phenyl]boronic acid
SMILES CN(C)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 227°C
Quantity 1 g
Formula Weight 165
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-(Dimethylamino)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.