missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)phenethyl alcohol, 99%

Product Code. 11310800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11310800 1 g Glass bottle 1g
11995271 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310800 Supplier Thermo Scientific Acros Supplier No. 210930010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50438-75-0 | C10H15NO | 165.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50438-75-0
InChI Key CDTPAAZQBPSVGS-UHFFFAOYSA-N
Synonym 2-4-dimethylamino phenyl ethanol, 4-dimethylamino phenethyl alcohol, ccris 987, 4-dimethylamino benzeneethanol, 2-4-dimetylaminophenyl ethanol, unii-84gfy3663n, 2-4-dimethylaminophenyl ethanol, benzeneethanol, 4-dimethylamino, p-n,n-dimethylaminophenethanol
PubChem CID 93252
IUPAC Name 2-[4-(dimethylamino)phenyl]ethanol
SMILES CN(C)C1=CC=C(C=C1)CCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 53.9°C to 55.1°C
Color White to Brown
Boiling Point 116.0°C (0.1 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H15NO
Linear Formula (CH3)2NC6H4CH2CH2OH
Quantity 1 g
Molecular Weight (g/mol) 165.23
Formula Weight 165.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Dimethylamino)phenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Dimethylamino)phenethyl alcohol, 99%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.