missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)tetrahydro-2H-pyran-4-carbonitrile, 97%, Thermo Scientific™

Product Code. 11346885
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11346885 5 g Amber Glass Bottle 5g
10326814 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346885 Supplier Thermo Scientific Maybridge Supplier No. MO00628DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 176445-77-5
Molecular Formula C8H14N2O
Molecular Weight (g/mol) 154.21
MDL Number MFCD09065026
InChI Key FDGOISRHLNMQRE-UHFFFAOYSA-N
Synonym 4-dimethylamino tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino oxane-4-carbonitrile, 4-cyano-4-dimethylamino-tetrahydro-4h-pyrane, 2h-pyran-4-carbonitrile,4-dimethylamino tetrahydro, 4-dimethylamino-tetrahydro-2h-pyran-4-carbonitrile, 4-dimethylamino-2h-3,4,5,6-tetrahydropyran-4-carbonitrile
PubChem CID 10511078
IUPAC Name 4-(dimethylamino)oxane-4-carbonitrile
SMILES CN(C)C1(CCOCC1)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 4-(Dimethylamino)tetrahydro-2H-pyran-4-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.