missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminoazobenzene-4'-sulfonyl chloride, 98+%

Product Code. 11322346
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322346 500 mg 500mg
11332346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322346 Supplier Thermo Scientific Alfa Aesar Supplier No. L06057.ME
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 56512-49-3 | C14H14ClN3O2S | 323.80 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56512-49-3
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00007444
InChI Key VTVWTPGLLAELLI-UHFFFAOYSA-N
Synonym dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride
PubChem CID 91660
IUPAC Name 4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonyl chloride
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point ∽185°C (decomposition)
Quantity 500 mg
UN Number UN3261
Beilstein 3064095
Sensitivity Moisture sensitive
Formula Weight 323.8
Percent Purity ≥98%
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-sulfonyl chloride
compliance-icons
Product Identifier
  • 4-Dimethylaminoazobenzene-4'-sulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 4-Dimethylaminoazobenzene-4'-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.