missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzhydrazide, 98+%

Product Code. 11424734
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424734 10 g 10g
11434734 50 g 50g
11444734 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424734 Supplier Thermo Scientific Alfa Aesar Supplier No. A13723.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19353-92-5 | C9H13N3O | 179.223 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19353-92-5
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD00014761
InChI Key HITIGLAGJBMISF-UHFFFAOYSA-N
Synonym 4-dimethylamino benzohydrazide, 4-dimethylamino benzhydrazide, 4-dimethylaminobenzhydrazide, p-dimethylamino benzohydrazide, 4-n,n-dimethylamino benzhydrazide, dimethyl 4-???phenyl amine, 4me2nphcon2, acmc-1bqxb, maybridge1_002636, 4-dimethylaminobenzoic acid, hydrazide
PubChem CID 88019
IUPAC Name 4-(dimethylamino)benzohydrazide
SMILES CN(C)C1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 174°C to 175°C
Quantity 10 g
Beilstein 2210856
Formula Weight 179.22
Percent Purity ≥98%
Chemical Name or Material 4-Dimethylaminobenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.