missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobenzylidenerhodanine, Extra Pure, SLR, Fisher Chemical

Product Code. 10050120 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass Jar
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10050120 5 g Glass Jar 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10050120 Supplier Fisher Chemical Supplier No. D/3650/44

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 536-17-4
Melting Point 285°C
Color Red
Packaging Glass Jar
Molecular Formula C12H12N2OS2
MDL Number 64857
Quantity 5 g
Synonym unii-0ser53q7rt, p-dimethylaminobenzalrhodanine, 5-4-dimethylaminobenzylidene rhodanine, 0ser53q7rt, usaf pd-20, 4-dimethylaminobenzylidenerhodanine, 4-thiazolidinone, 5-4-dimethylamino phenyl methylene-2-thioxo, 5-p-dimethylaminobenzal rhodanine, p-dimethylaminobenzylidene rhodamine, p-dimethylamino benzal-5-rhodanine
InChI Key JJRVRELEASDUMY-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)C=C2C(=O)NC(=S)S2
IUPAC Name 5-[[4-(dimethylamino)phenyl]methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol) 264.361
PubChem CID 2723826
Formula Weight 264.35g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.