missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminobutyric acid hydrochloride, 98%

Product Code. p-7023251
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11427957

Brand: Thermo Scientific Alfa Aesar A16610.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69954-66-1 | C6H14ClNO2 | 167.633 g/mol

4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobutyric acid hydrochloride is used as pharmaceutical intermediates.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas. Incompatible with water and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69954-66-1
Molecular Formula C6H14ClNO2
Molecular Weight (g/mol) 167.633
MDL Number MFCD00012613
InChI Key RDTALXUBMCLWBB-UHFFFAOYSA-N
Synonym 4-dimethylamino butanoic acid hydrochloride, 4-dimethylaminobutyric acid hydrochloride, 4-dimethylamino butyric acid hydrochloride, butanoic acid, 4-dimethylamino-, hydrochloride, 4-dimethylamino butanoic acid hcl, 4-dimethylamino butyric acid hcl salt, 4-dimethylamino-butyric acid hydrochloride, 4-dimethylamino-butyric acid hydrochloride salt
PubChem CID 12691640
IUPAC Name 4-(dimethylamino)butanoic acid;hydrochloride
SMILES CN(C)CCCC(=O)O.Cl

Specifications

Melting Point 153°C to 155°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 167.64
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobutyric acid hydrochloride
compliance-icons
Product Identifier
  • 4-Dimethylaminobutyric acid, hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Dimethylaminobutyric acid, hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.