missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminophenyl isothiocyanate, 97%

Product Code. 11312836
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312836 2 g 2g
11322836 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312836 Supplier Thermo Scientific Alfa Aesar Supplier No. L09011.04
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2131-64-8 | C9H10N2S | 178.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2131-64-8
Molecular Formula C9H10N2S
Molecular Weight (g/mol) 178.253
MDL Number MFCD00041093
InChI Key HRDJPEMAGYHSJR-UHFFFAOYSA-N
PubChem CID 75047
IUPAC Name 4-isothiocyanato-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 64°C to 70°C
Boiling Point 152°C (6 mmHg)
Odor Irritating
Quantity 2 g
UN Number UN2928
Beilstein 388270
Sensitivity Moisture sensitive
Synonym 4-dimethylamino phenyl isothiocyanate, 4-dimethylaminophenyl isothiocyanate, 4-n,n-dimethylamino phenyl isothiocyanate, p-dimethylamino phenyl isothiocyanate, benzenamine, 4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylaminophenyl ester, p-n-dimethylamino phenyl isothiocyanate, 4-dimethylamino phenylisothiocyanate, benzenamine,4-isothiocyanato-n,n-dimethyl, isothiocyanic acid, p-dimethylamino phenyl ester
Formula Weight 178.25
Percent Purity 97%
Chemical Name or Material 4-Dimethylaminophenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.