missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine, 99%

Product Code. 10091630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10091630 25 g Glass bottle 25g
10504081 5 g Glass bottle 5g
10133480 100 g Glass bottle 100g
10409633 500 g Glass bottle 500g
11916301 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10091630 Supplier Thermo Scientific Acros Supplier No. 148270250
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1122-58-3 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-58-3
MDL Number MFCD00006418
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
PubChem CID 14284
IUPAC Name N,N-dimethylpyridin-4-amine
SMILES CN(C)C1=CC=NC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 110.0°C to 113.0°C
Boiling Point 162.0°C (50.0 mmHg)
Flash Point 124°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H10N2
Linear Formula (C5H4N)N(CH3)2
Quantity 25 g
Beilstein 22, II, 341
Fieser 03,118; 09,178; 10,155; 12,199; 14,132; 15,131; 16,142
Merck Index 15, 3433
Solubility Information Solubility in water: 76g/L (25°C). Other solubilities: soluble in ether, slightly soluble in alcohol, benzene and, chloroform
Molecular Weight (g/mol) 122.17
Formula Weight 122.17
Vapor Pressure Negligible
Percent Purity 99%
Chemical Name or Material 4-Dimethylaminopyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Dimethylaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.