missing translation for 'onlineSavingsMsg'
Learn More

4-Dimethylaminopyridine (Peptide Synthesis), Fisher BioReagents

Product Code. 11413713
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11413713 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11413713 Supplier Fisher BioReagents Supplier No. BP596100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
4-Dimethylaminopyridine is a catalyst for several acylation reactions, especially in oligonucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Dimethylaminopyridine
CAS 1122-58-3
Melting Point 110°C
Color White
pH 9.7
Boiling Point 162°C
Assay Percent Range ≥99 %
Packaging Glass Bottle
Molecular Formula C7H10N2
Quantity 100 g
Merck Index 15, 3433
Synonym 4-dimethylaminopyridine, dmap, 4-dimethylamino pyridine, 4-pyridinamine, n,n-dimethyl, n,n-dimethyl-4-pyridinamine, p-dimethylaminopyridine, pyridine, 4-dimethylamino, ccris 6176, dimethylpyridin-4-ylamine
InChI Key VHYFNPMBLIVWCW-UHFFFAOYSA-N
SMILES CN(C)C1=CC=NC=C1
IUPAC Name N,N-dimethylpyridin-4-amine
Molecular Weight (g/mol) 122.171
PubChem CID 14284
Formula Weight 122.17
Percent Purity ≥99%
Grade Peptide Synthesis
Physical Form Solid
Chemical Name or Material 4-Dimethylaminopyridine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Dimethylaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.