missing translation for 'onlineSavingsMsg'
Learn More

4-(Diphenylamino)benzaldehyde, 98%

Product Code. 15452289
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15452289 25 g 25g
15432289 1 g 1g
15442289 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15452289 Supplier Thermo Scientific Alfa Aesar Supplier No. H64887.14
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4181-05-9 | C19H15NO | 273.34 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.34
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

Specifications

Melting Point 129°C to 133°C (literature)
Color Yellow
Odor Odorless
Quantity 25 g
Beilstein 2732795
Sensitivity Air sensitive
Solubility Information Soluble in methanol.
Formula Weight 273.33
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-(Diphenylamino)benzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.