missing translation for 'onlineSavingsMsg'
Learn More

4-Diphenylaminobenzaldehyde, 98%, Thermo Scientific Chemicals

Product Code. 10561954 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10561954 5 g Glass bottle 5g
10503325 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10561954 Supplier Thermo Scientific Acros Supplier No. 391180050
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4181-05-9 | Molecular Formula: C19H15NO | Molecular Weight: 273.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 4181-05-9
Molecular Formula C19H15NO
Molecular Weight (g/mol) 273.33
MDL Number MFCD00145131
InChI Key UESSERYYFWCTBU-UHFFFAOYSA-N
Synonym 4-diphenylamino benzaldehyde, 4-n,n-diphenylamino benzaldehyde, 4-diphenylaminobenzaldehyde, 4-diphenylamino-benzaldehyde, benzaldehyde, 4-diphenylamino, 4-formyltriphenylamine, p-formyltriphenylamine, 4-n-phenylanilino benzaldehyde, p-diphenylaminobenzaldehyde, 4-n,n-diphenylamino-benzaldehyde
PubChem CID 77846
IUPAC Name 4-(N-phenylanilino)benzaldehyde
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)C=O

Spécification

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129°C to 133°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)2NC6H4C(=O)H
Quantity 5 g
Formula Weight 273.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Diphenylaminobenzaldehyde
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 4-Diphenylaminobenzaldehyde
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.