missing translation for 'onlineSavingsMsg'
Learn More

4-Dodecylbenzenesulfonic Acid Potassium Salt, TRC

Product Code. 30180303
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180303 1 g 1g
30180409 10 g 10g
30179781 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180303 Supplier Toronto Research Chemicals Supplier No. TRCD4945601G
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14564-74-0
Molecular Formula C18H29KO3S
Molecular Weight (g/mol) 364.58
Synonym p-Dodecylbenzenesulfonic Acid Potassium Salt
SMILES [K+].CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C18H30O3S.K/c1-2-3-4-5-6-7-8-9-10-11-12-17-13-15-18(16-14-17)22(19,20)21;/h13-16H,2-12H2,1H3,(H,19,20,21);/q;+1/p-1
Chemical Name or Material 4-Dodecylbenzenesulfonic Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.