missing translation for 'onlineSavingsMsg'
Learn More

4’-Epi Lamivudine, TRC

Product Code. 30370045
Change view
Click to view available options
Quantity:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30370045 10 mg
30376891 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30370045 Supplier Toronto Research Chemicals Supplier No. TRCE58852510MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139757-68-9
Molecular Formula C8 H11 N3 O3 S
Molecular Weight (g/mol) 229.26
Synonym 2(1H)-Pyrimidinone, 4-amino-1-[(2R,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, 2(1H)-Pyrimidinone, 4-amino-1-[2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-, (2R-trans)- (9CI), 1,3-Oxathiolane, 2(1H)-pyrimidinone deriv., 4-Amino-1-[(2R,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]-2(1H)-pyrimidinone, trans(+)-2-Hydroxymethyl-5-(cytosin-1-yl)-1,3-oxathiolane, 4'-Epi Lamivudine, 4'-epi-Lamivudine, 4'-Epilamivudine, trans-Lamivudine, 4-Amino-1-[(2R,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2(1H)-one, (2'R-trans)-Lamivudine
IUPAC Name 4-amino-1-[(2R,5R)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl]pyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1)[C@H]2CS[C@H](CO)O2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H11N3O3S/c9-5-1-2-11(8(13)10-5)6-4-15-7(3-12)14-6/h1-2,6-7,12H,3-4H2,(H2,9,10,13)/t6-,7-/m1/s1
Formula Weight 229.0521
Chemical Name or Material Trans-Lamivudine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.