missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-nitropyridine hydrochloride, 97%, Thermo Scientific™

Product Code. 11313676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313676 25 g 25g
11303676 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313676 Supplier Thermo Scientific Alfa Aesar Supplier No. L14743.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Ethoxy-3-nitropyridine hydrochloride acts as an intermediate used in organic synthesis. It is also used in the preparation of 3,4-diaminopyridine.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive.Incompatible with oxidizing agent and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 94602-04-7
Molecular Formula C7H9ClN2O3
Molecular Weight (g/mol) 204.61
MDL Number MFCD01075663
InChI Key HNMWMLXQJBWURC-UHFFFAOYSA-N
Synonym 4-ethoxy-3-nitropyridine hydrochloride, 3-nitro-4-ethoxypyridine hydrochloride, 4-ethoxy-3-nitro-pyridine hydrochloride, 4-ethoxy-3-nitropyridine hydrogen chloride, pyridine, 4-ethoxy-3-nitro-, monohydrochloride, 4-ethoxy-3-nitropyridinehydrochloride, pubchem6674, labotest-bb lt00454779, 4-ethoxy-3-nitropyridine hcl, ksc496g1h
PubChem CID 12407497
IUPAC Name 4-ethoxy-3-nitropyridine;hydrochloride
SMILES CCOC1=C(C=NC=C1)[N+](=O)[O-].Cl

Spezifikation

Melting Point ∼270°C
Quantity 25 g
Beilstein 3712456
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 204.61
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-3-nitropyridine hydrochloride
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.