missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15485365
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485365 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15485365 Supplier Thermo Scientific Alfa Aesar Supplier No. H58484.AD
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 131379-16-3 | C9H9IO2Zn | 341.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Ethoxycarbonyl)phenylzinc iodide is used as pharmaceutical intermediates.

Solubility
Reacts with water.

Notes
Store at 4°C. Air Sensitive. Store under dry inert gas. Store away from air. Inco
TRUSTED_SUSTAINABILITY

Specifications

CAS 131379-16-3
Density 1.023
Molecular Formula C9H9IO2Zn
MDL Number MFCD00671982
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym 4-ethoxycarbonylphenylzinc iodide, 4-ethoxycarbonyl phenylzinc iodide, 4-ethoxycarbonyl phenyl iodo zinc, 4-ethoxycarbonyl phenyl zinc ii iodide, 4-ethoxycarbonyl phenylzinc iodide solution, 4-ethoxycarbonylphenylzinc iodide 0.5 m in tetrahydrofuran, 4-ethoxycarbonylphenylzinc iodide, 0.5m in tetrahydrofuran, 4-ethoxycarbonyl phenylzinc iodide solution, 0.5 m in thf, 4-ethoxycarbonyl phenylzinc iodide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key DVAYVJGYZRYJEV-UHFFFAOYSA-M
SMILES [Zn+]I.CCOC(=O)C1=CC=[C-]C=C1
IUPAC Name 4-(ethoxycarbonyl)benzen-1-ide; iodozincylium
Molecular Weight (g/mol) 341.45
PubChem CID 5107659
Formula Weight 341.45
Chemical Name or Material 4-(Ethoxycarbonyl)phenylzinc iodide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.