missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)butylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15415345
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15415345 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415345 Supplier Thermo Scientific Alfa Aesar Supplier No. H58248.AD
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 265330-98-1 | C7H13BrO2Zn | 274.463 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Reacts with water.

Notes
Store at 4°C. Incompatible with oxidizing agents, acid chlorides, acids. Store under dry inert gas. Air Sensitive. Store away from air.
TRUSTED_SUSTAINABILITY

Specifications

CAS 265330-98-1
Density 0.978
Molecular Formula C7H13BrO2Zn
Linear Formula CH3CH2O2C(CH2)4ZnBr
MDL Number MFCD00671978
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym 5-ethoxy-5-oxopentylzinc bromide solution, 5-ethoxy-5-oxopentylzinc bromide, bromozinc 1+ ,ethyl pentanoate, ethyl 5-bromozincio pentanoate, 5-ethoxy-5-oxopentylzinc bromide 0.5 m in tetrahydrofuran, 5-ethoxy-5-oxopentylzinc bromide solution, 0.5 m in thf, 4-ethoxycarbonyl butylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
Solubility Information Reacts with water.
InChI Key JJSSUZYKRQXNFR-UHFFFAOYSA-M
SMILES CCOC(=O)CCC[CH2-].[Zn+]Br
IUPAC Name bromozinc(1+);ethyl pentanoate
Molecular Weight (g/mol) 274.463
PubChem CID 3260042
Formula Weight 274.46
Chemical Name or Material 4-(Ethoxycarbonyl)butylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.