missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)phenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15455365
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455365 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15455365 Supplier Thermo Scientific Alfa Aesar Supplier No. H58477.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 131379-15-2
Density 1.023
Molecular Formula C9H9BrO2Zn
MDL Number MFCD05664208
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym zinc bromide 4-ethoxycarbonyl benzen-1-ide 1/1/1, zinc 2+ ion 4-ethoxycarbonyl benzen-1-ide bromide
InChI Key AXFMOPVOFKMPJN-UHFFFAOYSA-M
SMILES CCOC(=O)C1=CC=[C-]C=C1.[Zn+2].[Br-]
IUPAC Name zinc;ethyl benzoate;bromide
Molecular Weight (g/mol) 294.453
PubChem CID 76518832
Formula Weight 294.45
Chemical Name or Material 4-(Ethoxycarbonyl)phenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.