missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethoxycarbonyl)phenyl isothiocyanate, 97%

Product Code. 11313216
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313216 10 g 10g
11303216 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313216 Supplier Thermo Scientific Alfa Aesar Supplier No. L12522.09
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1205-06-7 | C10H9NO2S | 207.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-06-7
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
MDL Number MFCD00022058
InChI Key MLOJHUCMCKBDLV-UHFFFAOYSA-N
Synonym 4-ethoxycarbonylphenyl isothiocyanate, 4-ethoxycarbonylphenylisothiocyanate, benzoic acid, 4-isothiocyanato-, ethyl ester, 4-ethoxycarbonyl phenyl isothiocyanate, ethyl p-isothiocyanatobenzoate, 4-ethoxycarbonylphenyl-isothiocyanate, benzoic acid,4-isothiocyanato-, ethyl ester, acmc-1brcn, p-carbethoxyphenylisothiocyanate, #
PubChem CID 71003
IUPAC Name ethyl 4-isothiocyanatobenzoate
SMILES CCOC(=O)C1=CC=C(C=C1)N=C=S

Specifications

Melting Point 52°C to 56°C
Boiling Point 123°C to 127°C (1 mmHg)
Odor Irritating
Quantity 10 g
UN Number UN2923
Beilstein 2213400
Sensitivity Moisture sensitive
Formula Weight 207.25
Percent Purity 97%
Chemical Name or Material 4-(Ethoxycarbonyl)phenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.