missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxycarbonylphenylboronic acid, 97%

Product Code. 10009101 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
Product Code. Quantity Packaging unitSize
10009101 5 g Glass bottle 5g
10476462 1 g Glass bottle 1g
10155683 25 g Glass bottle 25g
3 options
This item is not returnable. View return policy

Product Code. 10009101

Brand: Thermo Scientific 367790050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4334-88-7 | C9H11BO4 | 193.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4334-88-7
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179441
InChI Key ZLNFACCFYUFTLD-UHFFFAOYSA-N
Synonym 4-ethoxycarbonylphenyl boronic acid, 4-ethoxycarbonyl phenylboronic acid, 4-ethoxycarbonyl phenyl boronic acid, 4-ethoxycarbonyl benzeneboronic acid, 4-ethoxycarbonylbenzeneboronic acid, ethyl 4-boronobenzoate, p-ethoxycarbonylphenylboronic acid, ethyl 4-dihydroxyboranyl benzoate, 4-carbethoxyphenylboronic acid, benzoic acid, 4-borono-, 1-ethyl ester
PubChem CID 2734350
SMILES CCOC(=O)C1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 184°C to 189°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
IUPAC Name [4-(ethoxycarbonyl)phenyl]boronic acid
Formula Weight 193.99
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Ethoxycarbonylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Ethoxycarbonylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.