missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxymethylene-2-phenyloxazolin-5-one, 98+%

Product Code. 11419432
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11419432 1 g 1g
11429432 5 g 5g
11439432 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419432 Supplier Thermo Scientific Alfa Aesar Supplier No. A10155.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15646-46-5 | C12H11NO3 | 217.224 g/mol

4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Ethoxymethylene-2-phenyloxazolin-5-one acts as sensitizing agent. It is also used as pharmaceutical intermediates.

Solubility
Soluble in methanol (50 mg/ml), and water (partly).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15646-46-5
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.224
MDL Number MFCD00003204
InChI Key SJHPCNCNNSSLPL-NTMALXAHSA-N
Synonym oxazolone, oxazolone sensitizing agent, 5 4h-oxazolone, 4-ethoxymethylene-2-phenyl, unii-78il0j7n72, 4-ethoxymethylene-2-phenyl-2-oxazolin-5-one, 4-ethoxymethylene-2-phenyloxazolin-5-one, 4-ethoxymethylene-2-phenyl-2-oxazoline-5-one, 4z-4-ethoxymethylidene-2-phenyl-1,3-oxazol-5-one, 2-oxazolin-5-one, 4-ethoxymethylene-2-phenyl, 4-ethoxymethylene-2-phenyloxazolone
PubChem CID 1712094
IUPAC Name (4Z)-4-(ethoxymethylidene)-2-phenyl-1,3-oxazol-5-one
SMILES CCOC=C1C(=O)OC(=N1)C2=CC=CC=C2

Specifications

Melting Point 93°C to 97°C
Quantity 1 g
Beilstein 163055
Solubility Information Soluble in methanol (50mg/ml),and water (partly).
Formula Weight 217.22
Percent Purity ≥98%
Chemical Name or Material 4-Ethoxymethylene-2-phenyloxazolin-5-one

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.