missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylamino)benzoic acid, 97%, Thermo Scientific™

Product Code. 10712273
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10712273 250 mg 250mg
10512462 1 g 1g
10441854 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10712273 Supplier Thermo Scientific Maybridge Supplier No. BTB10315CB
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7409-09-8
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00102118
InChI Key SWXFMMWYVSYQGF-UHFFFAOYSA-N
Synonym 4-ethylamino benzoic acid, 4-ethylaminobenzoic acid, benzoic acid,4-ethylamino, maybridge1_002225, p-athylaminobenzoesaure, swxfmmwyvsyqgf-uhfffaoysa-n
PubChem CID 2775249

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula CH3CH2NHC6H4CO2H
Quantity 250 mg
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material 4-(Ethylamino)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.