missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11455787
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11455787 10 g 10g
11445787 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455787 Supplier Thermo Scientific Alfa Aesar Supplier No. B24353.09
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18856-63-8 | Molecular Formula: C9H9NS | Molecular Weight: 163.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18856-63-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.24
MDL Number MFCD00041101
InChI Key XXLWCGRHJFEMQH-UHFFFAOYSA-N
Synonym 4-ethylphenyl isothiocyanate, 1-ethyl-4-isothiocyanato-benzene, benzene,1-ethyl-4-isothiocyanato, 4-ethylbenzenisothiocyanate, acmc-20amit, 2-p-isothio-cyanatophenyl ethane, #
PubChem CID 140438
IUPAC Name 1-ethyl-4-isothiocyanatobenzene
SMILES CCC1=CC=C(C=C1)N=C=S

Specifications

Density 1.075
Boiling Point 245°C
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.6205
Quantity 10 g
UN Number UN2922
Beilstein 2802384
Sensitivity Moisture sensitive
Formula Weight 163.24
Percent Purity 97%
Chemical Name or Material 4-Ethylphenyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.