missing translation for 'onlineSavingsMsg'
Learn More

4-(Ethylsulfurate Sulfonyl)aniline, TRC

Product Code. 30387356
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387356 1 g
30382399 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387356 Supplier Toronto Research Chemicals Supplier No. TRCE9260001G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2494-89-5
Molecular Formula C8H11NO6S2
Molecular Weight (g/mol) 281.31
Synonym 1-Aminobenzene-4-(2-sulfatoethyl)sulfone, 1-Aminobenzene-4-beta-sulfatoethylsulfone, 2-(4-Aminobenzenesulfonyl)ethyl sulfate, 2-(4-Aminophenylsulfonyl)ethyl bisulfate, 2-[(4-Aminophenyl)sulfonyl]ethyl hydrogen sulfate, 2-[(p-Aminophenyl)sulfonyl]ethyl hydrogen sulfate, 4-(2-Sulfatoethylsulfonyl)aniline, 4-(beta-Sulfatoethylsulfonyl)aniline, 4-Amino-1-beta-sulfatoethylsulfonylbenzene, 4-Aminophenyl 2-sulfatoethyl sulfone, 4-Aminophenyl beta-hydroxyethyl sulfone sulfate ester, 4-Aminophenyl beta-sulfatoethyl sulfone, 4-[(beta-Sulfooxyethyl)sulfonyl]aniline, Para Base Sulfate, Sodium 4-(beta-sulphatoethyl)sulfonylaniline, [2-[(4-Aminobenzene)sulfonyl]ethoxy]sulfonic acid, p-(2-Sulfatoethylsulfonyl)aniline, p-(beta-Sulfatoethylsulfonyl)aniline, p-Aminophenyl 2-sulfatoethyl sulfone, p-Aminophenyl beta-sulfatoethyl sulfone
SMILES Nc1ccc(cc1)S(=O)(=O)CCOS(=O)(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H11NO6S2/c9-7-1-3-8(4-2-7)16(10,11)6-5-15-17(12,13)14/h1-4H,5-6,9H2,(H,12,13,14)
Formula Weight 281.0028
Chemical Name or Material 4-(Ethylsulfurate Sulfonyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.