Learn More
4-Ethynylbenzeneboronic acid pinacol ester, 95%
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Especificaciones
Especificaciones
| Quantity | 1 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 228.1 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-Ethynylbenzeneboronic acid pinacol ester |
RUO – Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.