missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoro-2'-hydroxyacetophenone, 98%

Product Code. 11408676
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408676 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11408676 Supplier Thermo Scientific Alfa Aesar Supplier No. A16030.22
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1481-27-2 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1481-27-2
Melting Point 29°C to 34°C
Boiling Point 65°C (2 mmHg)
Flash Point 98°C
Molecular Formula C8H7FO2
MDL Number MFCD00143203
Quantity 100 g
Beilstein 2249799
Synonym 4 '-fluoro-2 '-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethanone, 4-fluoro-2-hydroxyacetophenone, 1-4-fluoro-2-hydroxyphenyl ethan-1-one, ethanone, 1-4-fluoro-2-hydroxyphenyl
InChI Key HLTBTUXAMVOKIH-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(F)C=C1O
IUPAC Name 1-(4-fluoro-2-hydroxyphenyl)ethanone
Molecular Weight (g/mol) 154.14
PubChem CID 2737326
Formula Weight 154.14
Percent Purity 98%
Chemical Name or Material 4 '-Fluoro-2 '-hydroxyacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.