missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-nitroaniline, 99%

Product Code. 10125010 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
Product Code. Quantity Packaging unitSize
10125010 10 g Glass bottle 10g
10592251 100 g Glass bottle 100g
2 options
This item is not returnable. View return policy

Product Code. 10125010

Brand: Thermo Scientific Acros 233070100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 364-78-3 | C6H5FN2O2 | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-78-3
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007830
InChI Key PUGDHSSOXPHLPT-UHFFFAOYSA-N
Synonym 2-nitro-4-fluoroaniline, 4-fluoro-2-nitrophenylamine, 4-fluoro-2-nitrobenzeneamine, 4-fluoro-2-nitrobenzenamine, benzenamine, 4-fluoro-2-nitro, 2-amino-5-fluoronitrobenzene, 4-fluoro-2-nitro-aniline, 4-fluoro-2-nitro-phenylamine, 4-fluor-2-nitroanilin, pubchem3421
PubChem CID 67769
IUPAC Name 4-fluoro-2-nitroaniline
SMILES NC1=CC=C(F)C=C1[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 92°C to 95°C
Color Orange to Brown
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 10 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in polar organic solvents,insoluble in non-polar organic solvents,approx. 12% soluble in alcohol (15°C)
Formula Weight 156.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Fluoro-2-nitroaniline, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.