missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitroaniline, 98%

Product Code. 11408316
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11408316

Brand: Thermo Scientific Alfa Aesar A15371.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 364-76-1 | C6H5FN2O2 | 156.12 g/mol

4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitroaniline is used in the preparation of commercial hair dyes. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Incompatible with Oxidizing agents, Acids, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-76-1
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007833
InChI Key LLIOADBCFIXIEU-UHFFFAOYSA-N
Synonym 3-nitro-4-fluoroaniline, 4-fluoro-3-nitrobenzenamine, benzenamine, 4-fluoro-3-nitro, aniline, 4-fluoro-3-nitro, 4-fluoro-3-nitro-aniline, 5-amino-2-fluoronitrobenzene, 4-fluoro-3-nitro-phenylamine, 3-nitro-4-fluoroanailine, 3-nitro-4-fluoro aniline, 4-fluoro-3-nitro aniline
PubChem CID 67768
ChEBI CHEBI:48642
IUPAC Name 4-fluoro-3-nitroaniline
SMILES NC1=CC=C(F)C(=C1)[N+]([O-])=O

Specifications

Melting Point 94°C to 98°C
Flash Point 91°C (195°F)
Quantity 25 g
Beilstein 2210199
Solubility Information Insoluble in water.
Formula Weight 156.12
Percent Purity 98%
Chemical Name or Material 4-Fluoro-3-nitroaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.