missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15457776
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15457776 250 mg 250mg
15447776 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457776 Supplier Thermo Scientific Alfa Aesar Supplier No. H62720.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-nitrobenzeneboronic acid is generally used as a intermediate in pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1218791-09-3
Molecular Formula C12H15BFNO4
MDL Number MFCD12546515
Synonym 2-3-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-fluoro-4-nitrophenylboronic acid pinacol ester, 3-fluoro-4-nitrophenylboronic acid,pinacol ester, 3-fluoro-4-nitrophenylboronic acid, pinacol ester, 2-3-fluoro-4-nitrophenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane
PubChem CID 46739531

Specifications

Melting Point 64°C to 65°C
Color Yellow
Quantity 250 mg
Beilstein 20289173
Solubility Information Slightly soluble in water.
InChI Key MOKWUYKZXFBLKR-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)[N+](=O)[O-])F
IUPAC Name 2-(3-fluoro-4-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 267.063
Formula Weight 267.06
Percent Purity 96%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluoro-3-nitrobenzeneboronic acid pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.