missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-nitrobenzonitrile, 97%

Product Code. 15415117
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15415117

Brand: Thermo Scientific Alfa Aesar H32391.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1009-35-4 | C7H3FN2O2 | 166.111 g/mol

4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluoro-3-nitrobenzonitrile, is used as a pharmaceutical intermediate. It is also used as a synthetic and fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-35-4
Molecular Formula C7H3FN2O2
Molecular Weight (g/mol) 166.111
MDL Number MFCD01632197
InChI Key LKOWKPGBAZVHOF-UHFFFAOYSA-N
Synonym 3-nitro-4-fluorobenzonitrile, 4-fluoro-3-nitro-benzonitrile, 5-cyano-2-fluoronitrobenzene, 4-cyano-1-fluoro-2-nitrobenzene, benzonitrile, 4-fluoro-3-nitro, 4-fluoro-3-nitrobenzenecarbonitrile, 5-cyano-5-fluoro-1-nitrobenzene, pubchem4796, acmc-1c2vd, 4-fluro-3-nitrobenzonitrile
PubChem CID 2774654
IUPAC Name 4-fluoro-3-nitrobenzonitrile
SMILES C1=CC(=C(C=C1C#N)[N+](=O)[O-])F

Specifications

Melting Point 90°C to 94°C
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 166.11
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-nitrobenzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.