missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethyl)phenacyl bromide, Thermo Scientific™

Product Code. 10139679
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10139679 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10139679 Supplier Thermo Scientific Maybridge Supplier No. MAY00240DA
 Restricted Item
Estimated Shipment: 18-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 537050-14-9
Packaging Amber Glass Bottle
Molecular Formula C9H5BrF4O
Quantity 1 g
Synonym 4-fluoro-3-trifluoromethyl phenacyl bromide, 2-bromo-1-4-fluoro-3-trifluoromethyl phenyl ethanone, 4-fluoro-3-trifluoromethyl phenacylbromid, 4-fluoro-3-trifluoromethyl phenacyl bromid, 2-bromo-1-4-fluoro-3-trifluoromethyl phenyl ethan-1-one, 2-bromo-4'-fluoro-3'-trifluoromethyl acetophenone, 4-fluoro-3-trifluoromethyl phenacylbromide, 2-bromo-4'-fluoro-3'-trifluoromethylacetophenone, acmc-209l9v
InChI Key BUNOSKWLVXNWEH-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(=O)CBr)C(F)(F)F)F
IUPAC Name 2-bromo-1-[4-fluoro-3-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol) 285.036
PubChem CID 2778787
Formula Weight 285.04
Percent Purity 95%
Chemical Name or Material 4-Fluoro-3-(trifluoromethyl)phenacyl bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.