missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroacetophenone, 99%

Product Code. 11491688
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491688 500 g 500g
11471688 25 g 25g
11481688 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491688 Supplier Thermo Scientific Alfa Aesar Supplier No. A13950.36
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 403-42-9 | C8H7FO | 138.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-42-9
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000354
InChI Key ZDPAWHACYDRYIW-UHFFFAOYSA-N
Synonym 4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
PubChem CID 9828
IUPAC Name 1-(4-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C=C1

Specifications

Melting Point 4°C
Density 1.141
Boiling Point 194°C to 196°C
Flash Point 71°C (159°F)
Refractive Index 1.511
Quantity 500 g
Beilstein 386013
Formula Weight 138.14
Percent Purity 99%
Chemical Name or Material 4'-Fluoroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.