missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluoroacetophenone, 99%

Product Code. 10676492 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10676492 25 mL Glass bottle 25mL
10686492 100 mL Glass bottle 100mL
10512544 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10676492 Supplier Thermo Scientific Acros Supplier No. 119260250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 403-42-9 | C8H7FO | 138.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-42-9
Molecular Formula C8H7FO
Molecular Weight (g/mol) 138.14
MDL Number MFCD00000354
InChI Key ZDPAWHACYDRYIW-UHFFFAOYSA-N
Synonym 4'-fluoroacetophenone, 4-fluoroacetophenone, 1-4-fluorophenyl ethanone, p-fluoroacetophenone, 1-4-fluorophenyl ethan-1-one, ethanone, 1-4-fluorophenyl, acetophenone, 4'-fluoro, 4 fluoroacetophenone, unii-tvq090602v, 1-acetyl-4-fluorobenzene
PubChem CID 9828
IUPAC Name 1-(4-fluorophenyl)ethanone
SMILES CC(=O)C1=CC=C(F)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 4°C
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 196°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4COCH3
Refractive Index 1.5090 to 1.512
Quantity 25 mL
Specific Gravity 1.13
Formula Weight 138.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4'-Fluoroacetophenone, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Fluoroacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.