missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoroaniline, TRC

Product Code. 30407350
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30407350 250 g
30405250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407350 Supplier Toronto Research Chemicals Supplier No. TRCF587650250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-40-4
Molecular Formula C6 H6 F N
Molecular Weight (g/mol) 111.12
Synonym Benzenamine, 4-fluoro- (9CI, ACI), 4-Fluorobenzenamine (ACI), Aniline, p-fluoro- (8CI), 1-Amino-4-fluorobenzene, 4-Fluoro-1-benzenamine, 4-Fluoroaniline, 4-Fluorophenylamine, NSC 579, p-Fluoroaniline, p-Fluorophenylamine
IUPAC Name 4-fluoroaniline
SMILES Nc1ccc(F)cc1

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H6FN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2
Formula Weight 111.0484
Chemical Name or Material 4-Fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.