missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzaldehyde, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Fluorobenzaldehyde |
| CAS | 459-57-4 |
| Molecular Formula | C7H5FO |
| Quantity | 5 g |
| Synonym | NSC 68095, p-Fluorobenzaldehyde, p-Fluorobenzenecarboxaldehyde |
| InChI Formula | InChI=1S/C7H5FO/c8-7-3-1-6(5-9)2-4-7/h1-5H |
| SMILES | Fc1ccc(C=O)cc1 |
| Molecular Weight (g/mol) | 124.11 |
| Formula Weight | 124.0324 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction