missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzenesulfonyl chloride, 98%

Product Code. 10397080 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10397080 100 g Glass bottle 100g
10152290 25 g Glass bottle 25g
10269533 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10397080 Supplier Thermo Scientific Supplier No. 119351000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 349-88-2 | C6H4ClFO2S | 194.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 349-88-2
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.60
MDL Number MFCD00007438
InChI Key BFXHJFKKRGVUMU-UHFFFAOYSA-N
Synonym 4-fluorobenzene-1-sulfonyl chloride, p-fluorobenzenesulfonyl chloride, 4-fluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 4-fluoro, 4-fluorophenylsulfonyl chloride, 4-fluorobenzenesulfonic acid chloride, 4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, p-fluoro, 4-fluorobenzensulfonyl chloride, 4-fluoro-benzenesulfonyl chloride
PubChem CID 9588
IUPAC Name 4-fluorobenzenesulfonyl chloride
SMILES FC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 29°C to 34°C
Color Brown to White
Boiling Point 95°C to 96°C (2.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula FC6H4SO2Cl
Quantity 100 g
Beilstein 11, 53
Formula Weight 194.61
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 4-Fluorobenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorobenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.