missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoyl Chloride, TRC

Product Code. 30391144
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391144 10 g 10g
30400497 50 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391144 Supplier Toronto Research Chemicals Supplier No. TRCF58835510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 403-43-0
Molecular Formula C7H4ClFO
Molecular Weight (g/mol) 158.56
Synonym 4-Fluorobenzoyl Chloride, 4-Fluorobenzenecarbonyl Chloride, 4-Fluorobenzoic Acid Chloride, 4-Fluorobenzoyl Chloride, NSC 88305, p-Fluorobenzoic Acid Chloride, p-Fluorobenzoyl Chloride
SMILES Fc1ccc(cc1)C(=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H4ClFO/c8-7(10)5-1-3-6(9)4-2-5/h1-4H
Formula Weight 157.9935
Chemical Name or Material 4-Fluorobenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.