missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluorobiphenyl-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15289646 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15289646 5 g 5g
15279646 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15289646 Supplier Thermo Scientific Acros Supplier No. 450320050
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 116748-66-4 | C12H8ClFO2S | 270.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116748-66-4
Molecular Formula C12H8ClFO2S
Molecular Weight (g/mol) 270.71
InChI Key CIDMHDJTWVMBIF-UHFFFAOYSA-N
Synonym 4'-fluorobiphenyl-4-sulfonyl chloride, 4-4-fluorophenyl benzenesulfonyl chloride, 4'-fluoro 1,1'-biphenyl-4-sulfonyl chloride, 4'-fluoro-biphenyl-4-sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-fluoro, 4'-fluoro-1,1'-biphenyl-4-sulfonyl chloride, acmc-20apun, 4'-fluorobiphenyl-4-sulfonylchloride, 4'-fluoro-4-biphenylsulfonyl chloride
PubChem CID 2794749
IUPAC Name 4-(4-fluorophenyl)benzenesulfonyl chloride
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)S(=O)(=O)Cl)F

Specifications

Melting Point 82.0°C to 84.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 270.71
Percent Purity 97%
Chemical Name or Material 4'-Fluorobiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 4'-Fluorobiphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.