missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenethyl Chloride, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 4-Fluorophenethyl Chloride |
| CAS | 332-43-4 |
| Molecular Formula | C8H8FCl |
| Quantity | 100 mg |
| Synonym | p-Fluorophenethyl Chloride, 1-(2-Chloroethyl)-4-fluorobenzene |
| InChI Formula | InChI=1S/C8H8ClF/c9-6-5-7-1-3-8(10)4-2-7/h1-4H,5-6H2 |
| SMILES | Fc1ccc(CCCl)cc1 |
| IUPAC Name | 1-(2-chloroethyl)-4-fluorobenzene |
| Molecular Weight (g/mol) | 158.6 |
| Formula Weight | 158.0299 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction