missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenethylamine, TRC

Product Code. 30399472
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399472 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399472 Supplier Toronto Research Chemicals Supplier No. TRCF593875500MG
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1583-88-6
Molecular Formula C8H10FN
Quantity 500 mg
Synonym 4-Fluorobenzeneethanamine (ACI), Phenethylamine, p-fluoro- (8CI), (p-Fluorophenyl)ethylamine, 2-(4-Fluorophenyl)ethan-1-amine, 2-(4-Fluorophenyl)ethanamine, 2-(4-Fluorophenyl)ethylamine, 2-(p-Fluorophenyl)ethylamine, 4-(2-Aminoethyl)fluorobenzene, 4-Fluorophenethylamine, N-(4-Fluorophenethyl)amine, p-Fluorophenethylamine
InChI Formula InChI=1S/C8H10FN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5-6,10H2
SMILES NCCc1ccc(F)cc1
Molecular Weight (g/mol) 139.17
Formula Weight 139.0797
Chemical Name or Material 4-Fluorophenethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.