missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenoxy-ethylbromide, 95%, Thermo Scientific™

Product Code. 10245220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10245220 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10245220 Supplier Thermo Scientific Acros Supplier No. 124160250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 332-48-9
CAS Min % 94.0
CAS Max % 100.0
Color Brown to Colorless
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8BrFO
Linear Formula FC6H4O(CH2)2Br
Refractive Index 1.5300 to 1.532
MDL Number MFCD00044739
Quantity 25 g
Synonym 1-2-bromoethoxy-4-fluorobenzene, 4-fluorophenoxy-ethylbromide, 2-bromoethyl 4-fluorophenyl ether, 4-fluorophenoxyethylbromide, 4-2-bromoethoxy fluorobenzene, 4-fluorophenoxyethyl bromide, beta-bromo-4-fluorophenetole, 2-4-fluorophenoxy ethyl bromide, benzene, 1-2-bromoethoxy-4-fluoro, p-fluorophenoxy ethylbromide
InChI Key JXSPKRUNMHMICQ-UHFFFAOYSA-N
SMILES FC1=CC=C(OCCBr)C=C1
IUPAC Name 1-(2-bromoethoxy)-4-fluorobenzene
Molecular Weight (g/mol) 219.05
PubChem CID 2064171
Formula Weight 219.05
Percent Purity 95%
Physical Form Liquid After Melting
Chemical Name or Material 4-Fluorophenoxy-ethylbromide, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenoxy-ethylbromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.