missing translation for 'onlineSavingsMsg'
Learn More

4'-Formyl(1,1'-biphenyl)-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10698535
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10698535 5 g 5g
10025222 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10698535 Supplier Thermo Scientific Maybridge Supplier No. MO01260DE
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 70916-98-2
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD03424613
InChI Key JCEAFZUEUSBESW-UHFFFAOYSA-N
Synonym 4-4-formylphenyl benzoic acid, 4'-formylbiphenyl-4-carboxylic acid, 4'-formyl-1,1'-biphenyl-4-carboxylic acid, 4'-formyl-biphenyl-4-carboxylic acid, 4-biphenyl-4'-formyl-carboxylic acid, 4'-formyl 1,1'-biphenyl-4-carboxylic acid, 1,1'-biphenyl-4-carboxylic acid, 4'-formyl, 4-biphenyl-4'-formyl-carboxylicacid, pubchem10282
PubChem CID 2794722
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 4'-formyl-[1,1'-biphenyl]-4-carboxylic acid
Formula Weight 226.23
Percent Purity 97%
Chemical Name or Material 4'-Formyl(1, 1'-biphenyl)-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.