missing translation for 'onlineSavingsMsg'
Learn More

4-Formylcinnamic acid, 98%, predominantly trans

Product Code. p-3778676 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10012390 1 g Glass bottle 1g
10012400 5 g Glass bottle 5g
10259443 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10012390 Supplier Thermo Scientific Acros Supplier No. 176590010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23359-08-2 | C10H8O3 | 176.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23359-08-2
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.17
MDL Number MFCD00006955
InChI Key LBOUHDMYVURTMA-AATRIKPKSA-N
Synonym 4-formylcinnamic acid, 3-4-formylphenyl acrylic acid, p-formylcinnamic acid, e-3-4-formylphenyl acrylic acid, 2e-3-4-formylphenyl prop-2-enoic acid, 4-2-carboxyvinyl benzaldehyde, p-formylcinnamicacid, 3-4-formylphenyl prop-2-enoic acid, 2e-3-4-formylphenyl acrylic acid, pubchem8244
PubChem CID 5357280
IUPAC Name (E)-3-(4-formylphenyl)prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 250°C to 255°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HCOC6H4CH=CHCO2H
Quantity 1 g
Formula Weight 176.17
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Formylcinnamic acid, Predominantly trans
Show More Show Less
compliance-icons
Product Identifier
  • 4-Formylcinnamic acid, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.