missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic Acid Hydrochloride Salt, TRC

Product Code. 30405498
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405498 25 g
30393565 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405498 Supplier Toronto Research Chemicals Supplier No. TRCG82130025G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42823-46-1
Molecular Formula C8 H9 N3 O2 . Cl H
Molecular Weight (g/mol) 215.64
Synonym 4-Guanidinobenzoic Acid Hydrochloride
IUPAC Name 4-carbamimidamidobenzoic acid;hydrochloride
SMILES Cl.NC(=N)Nc1ccc(cc1)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H9N3O2.ClH/c9-8(10)11-6-3-1-5(2-4-6)7(12)13;/h1-4H,(H,12,13)(H4,9,10,11);1H
Formula Weight 215.0462
Chemical Name or Material 4-Guanidinobenzoic Acid Hydrochloride Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.