missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobutanoic acid, TRC

Product Code. 30138105
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138105 500 mg 500mg
30135154 1 g 1g
30003734 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138105 Supplier Toronto Research Chemicals Supplier No. TRCG825100500MG
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 463-00-3
Molecular Formula C5 H11 N3 O2
Molecular Weight (g/mol) 145.16
Synonym Butanoic acid, 4-[(aminoiminomethyl)amino]-, Butyric acid, 4-guanidino- (6CI,8CI), 4-[(Aminoiminomethyl)amino]butanoic acid, 4-Guanidinobutyric acid, HL 6450, N-(4-Butanoic acid)guanidine, NSC 521717, γ-Guanidinebutyric acid, γ-Guanidinobutyric acid
IUPAC Name 4-guanidinobutanoic acid
SMILES NC(=N)NCCCC(=O)O

Specifications

InChI Formula InChI=1S/C5H11N3O2/c6-5(7)8-3-1-2-4(9)10/h1-3H2,(H,9,10)(H4,6,7,8)
Quantity 500 mg
Formula Weight 145.0851
Shipping Condition Room Temperature
Chemical Name or Material 4-Guanidinobutanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.