missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 4-HNE Competitive ELISA Kit
missing translation for 'orderingAttributeHoverText'
Quantity:
96 Tests
Beschreibung
4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, is an alpha, beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha, beta-unsaturated hydroxyalkenal formed in this process.
Spezifikation
Spezifikation
| Assay Range | 0.63 to 40 ng/mL |
| Assay Sensitivity | 0.38 ng/mL |
| Conjugate | Unlabeled |
| Product Type | ELISA |
| Sample Type | Cell Lysates, Plasma, Serum, Tissue Homogenates |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Interassay CV | <10 |
| Intraassay CV | <10 |
| Kit Contents | Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer |
| Label or Dye | Biotin |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Spot an opportunity for improvement?Share a Content Correction